Anti-5-HT Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-1126R-CY7EA 790 GBP
BOSSBS-1126R-CY7
Anti-5-HT Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Serotonin (pronounced) (5-hydroxytryptamine, or 5-HT) is a monoamine neurotransmitter synthesized in serotonergic neurons in the central nervous system (CNS) and enterochromaffin cells in the gastrointestinal tract of animals including humans. Serotonin is also found in many mushrooms and plants, including fruits and vegetables.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 5-HT
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR