Anti-c-phycocyanin Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-0697R-A488EA 790 GBP
BOSSBS-0697R-A488
Anti-c-phycocyanin Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
C-Phycocyanin (CPC), a pigment-protein complex from the light-harvesting phycobiliprotein family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: c-phycocyanin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR