Anti-TEP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-0632R-A647EA 805 GBP
BOSSBS-0632R-A647
Anti-TEP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
No Data Available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Thioster-containing protein 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR