3-(4-tert-Butylphenyl)isobutyraldehyde

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-tert-Butyl-alpha-methylhydrocinnamaldehyde3-(4-(tert-Butyl)phenyl)-2-methylpropanal3-(4-tert-Butylphenyl)-2-methylpropionaldehydeLilial

EHERCA10931620EA 89.7 GBP
EHERCA10931620
3-(4-tert-Butylphenyl)isobutyraldehyde
3-(4-tert-Butylphenyl)isobutyraldehyde
Formula: C₁₂H₂₀O
MW: 204.31 g/mol
Boiling Pt: 279 °C
Density: 0.946 g/cm³
Flash Pt: 79 °C
MDL Number: MFCD00047655
CAS Number: 80-54-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR