Poly(propylene carbonate), M.W. 50000 (GPC), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
389021-100G
389021-25G
SIAL389021-100GEA
476
GBP
SIAL389021-100G
SIAL389021-25G
Poly(propylene carbonate), M.W. 50000 (GPC), Sigma-Aldrich®
Poly(propylene carbonate)
Formula:
[CH(CH₃)CH₂OCO₂]n MW: ~50000 g/mol (average Mn) |
MDL Number:
MFCD00197919 CAS Number: 25511-85-7 |
Specification Test Results
Appearance (Color) | Colorless |
Appearance (Form) | Pellets |
Infrared spectrum | Conforms to Structure |
Assay (Propylene Carbonate) | 4,0 - 5,5% |
Storage Temperature | 2 to 8 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...