Calcium acetate 99.0-100.5% USP

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Calcium di(acetate)Acetic acid calcium saltCalcium acetate, United States Pharmacopeia (USP) Reference Standard

APPC141211.1211EA 113 GBP
APPC141211.1211 APPC141211.0416
Calcium acetate 99.0-100.5% USP
Calcium acetate
Formula: (H₃CCOO)₂Ca
MW: 158.17 g/mol
MDL Number: MFCD00150019
CAS Number: 62-54-4
EINECS: 200-540-9

Order Now

Specification Test Results

Assay (Compl.) (calc. a.a.s) 99,0-100,5%
Presentation White Small Crystals
Solubility Soluble in water 300 g/l at 20 °C, Hygroscopic
Termal Stability Thermal decomposition above 160 °C
pH (0,2 M sol.) 7,6
pH of 5% solution 6,3-9,6
Insoluble matter in H2O 0,1% Max.
Easily oxidable substances Passes Test
Chloride (Cl) 0,05% Max.
Phosphate(PO4) 0,005% Max.
Sulphate (SO4) 0,06% Max.
Residual solvents (Ph. Eur/USP) Passes Test
Fluoride (F) 0,005% Max.
Nitrate (NO3) Passes Test
Water (H2O) 7,0% Max.
Heavy metals (as Pb) 0,002% Max.
Aluminium (Al) 0,001% Max.
Arsenic (As) 0,0002% Max.
Barium (Ba) 0,005% Max.
Copper (Cu) 0,001% Max.
Iron (Fe) 0,005% Max.
Magnesium (Mg) 0,05% Max.
Nickel (Ni) 0,001% Max.
Lead (Pb) 0,001% Max.
Strontium (Sr) 0,05% Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR