Stain reagent, GelCode™
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GelCode®
PIER24592EA 619 GBP
PIER24592
Stain reagent, GelCode™
GelCode™ is a blue reagent offering a fast, sensitive method for SDS-PAGE gel staining.
  • Fast one-step staining protocol
  • More sensitive than standard Coomassie gel stains
  • Water Wash Enhancement™ Step increases staining sensitivity