Zinc protoporphyrin ≥95% (by TLC), Calbiochem®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Synonyms: Protoporphyrin IX zinc(II)Zinc (II) Protoporphyrin IXZinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate

691550-25EA 95 GBP
691550-25
Zinc protoporphyrin ≥95% (by TLC), Calbiochem®
Zinc protoporphyrin

A potent and selective inhibitor of heme oxygenase, the enzyme which generates carbon monoxide (CO) and biliverdin.


Inhibits soluble guanylyl cyclase and produces a time- and concentration-dependent inactivation of all three isoforms of nitric oxide synthase (IC50 = 800 nM, 4 µM, and 5 µM for nNOS, iNOS, and eNOS, respectively). Inhibits relaxation induced by acetylcholine, ANP, and VIP in isolated rat aorta. Blocks interleukin-1 and prevents induction of long-term potentiation. Also useful as an indicator of iron-deficient erythropoiesis. Does not cross the blood-brain barrier.


Caution: Protect from light.

Formula: C₃₄H₃₂N₄O₄Zn
MW: 626.034 g/mol
MDL Number: MFCD00011612
CAS Number: 15442-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR