Vitamin mixture, AIN-76-A
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0296009801EA 180 GBP
ICNA0296009801
Vitamin mixture, AIN-76-A
Animal Supplies Animal Feed
Basic component of animal diet especially for maintaining mice and rats colonies in the research laboratory.
Please see product datasheet for complete diet composition.
Delivery information: In powdered form.