Camera adapter
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMESP1030EA 70.5 GBP
SIMESP1030
Camera adapter
This adapter for digital camera is used to record destructive dry film thickness measurements.
  • Easy clamping of the adapter on the microscope
  • Pictures can be used for detailed reporting or as evidence
It can fit on the microscope and has a conical shaped inner part in non-glare plastic.
Delivery information: Supplied with a belt clip, in a leather pouch.