Blowers for fume hoods
Supplier: Labconco
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LBCP7071900-15419AEA 0 GBP
LBCP7071900-15419A
Blowers for fume hoods
Ventilators
These blowers are designed for fume hoods exhaust systems for low to moderate corrosive conditions.
  • Durable and easy to maintain
  • Blowers have low RPM for quiet operation
  • Phenolic coating on impeller and housing is corrosion resistant
  • Powder-coated base and weather cover provide long life in extreme weather
  • Forward curved impellers are balanced to ensure quiet operation and optimum air delivery
  • Flow-through ventent holes and louvers allows the air to circulate that cools the motor, thus prolonging motor life
  • Self-adjusting gravity belt tightener ensures consistent performance because belt is held taut by weight of motor
Sheaves are adjustable to allow for air volume adjustment. Pillow block bearings are factory lubricated to reduce maintenance and increase life.