Specific gravity hydrometers
Supplier: Brannan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRNN10/501/0EA 32.3 GBP
BRNN10/500/0 BRNN10/501/0
Specific gravity hydrometers
Hydrometers
Specific gravity hydrometers for low surface tension designed to compare the density of a liquid with that of water.
Certifications: ISO 650 and BS 718:1960.