2-Bromo-1-ethylpyridinium tetrafluoroborate 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate2-Bromo-1-ethyl-pyridinium tetrafluoroborate

ACRO439960050EA 84.8 GBP
ACRO439960050
2-Bromo-1-ethylpyridinium tetrafluoroborate 97%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formula: C₇H₉BBrF₄N
MW: 273.86 g/mol
Melting Pt: 100…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00060046
CAS Number: 878-23-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 108 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR