Benzoyl chloride 99%, AcroSeal®, pure

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Benzoic acid chloride

220271000.EA 48.1 GBP
220271000.
Benzoyl chloride 99%, AcroSeal®, pure
Benzoyl chloride
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 198 °C (760 mmHg)
Melting Pt: –1 °C
Density: 1.21 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
UN: 1736
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤40 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR