Methyl-4-iodobutanoate 95% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methyl 4-iodobutanoate, Methyl 4-iodobutyrate
426370010
426370050
ACRO426370010EA
81.8
GBP
ACRO426370010
ACRO426370050
Methyl-4-iodobutanoate 95% stabilised
Methyl-4-iodobutanoate
Appearance: Clear colorless to yellow Liquid
Formula:
C₅H₉IO₂ MW: 228.029 g/mol Boiling Pt: 80…83 °C (11 mmHg) Density: 1.689 g/cm³ Flash Pt: 99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00674066 CAS Number: 14273-85-9 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Stabilizer | Copper |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...