4-Nitrosalicylic acid 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxy-4-nitrobenzoic acid
ACRO425870250EA
244
GBP
ACRO425870250
4-Nitrosalicylic acid 97%
4-Nitrosalicylic acid
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Melting Pt: 235…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD00071505 CAS Number: 619-19-2 |
Specification Test Results
Appearance | White to yellow-grey to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 230 to 239 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...