Arachidyl alcohol
Supplier: Spectrum Chemical
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Eicosanol
SPCMA3850-5KGBLEA
0
GBP
SPCMA3850-5KGBL
Arachidyl alcohol
Arachidyl alcohol
Formula:
H₃C(CH₂)₁₈CH₂OH MW: 298.55 g/mol Boiling Pt: 309 °C (1013 hPa) Melting Pt: 71.5 °C Density: 0.88 g/cm³ (21 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002938 CAS Number: 629-96-9 EINECS: 211-119-4 |
Specification Test Results
Assay | 95% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...