trans-Cinnamic acid
Supplier: Spectrum Chemical
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(2E)-2-Phenyl-2-propenoic acid
, trans-Cinnamic acid
, Cinnamic acid (only trans)
SPCMCI120-25KGBLEA
0
GBP
SPCMCI120-25KGBL
trans-Cinnamic acid
trans-Cinnamic acid
Formula:
C₉H₈O₂ MW: 148.16 g/mol Boiling Pt: 300 °C (1013 hPa) Melting Pt: 131…134 °C Density: 1.25 g/cm³ (4 °C) |
MDL Number:
MFCD00004369 CAS Number: 140-10-3 EINECS: 205-398-1 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...