2-Amino-5-bromobenzothiazole 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H55270.03 H55270.06
H55270.03EA 39.6 GBP
H55270.03 H55270.06
2-Amino-5-bromobenzothiazole 97%
2-Amino-5-bromobenzothiazole
Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00577806
CAS Number: 20358-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR