5-Methoxy-1H-indole-3-carboxylic acid 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Methoxyindole-3-carboxylic acid
H64722.03
H64722.06
H64722.03EA
71
GBP
H64722.03
H64722.06
5-Methoxy-1H-indole-3-carboxylic acid 97%
5-Methoxy-1H-indole-3-carboxylic acid
Purity: 97%
CAS No.: 10242-01-0
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Appearance: Red
Beilstein Reference: 475711
MDL No.: MFCD03265451
Formula:
C10H9NO3 MW: 191.19 g/mol |
MDL Number:
MFCD03265451 CAS Number: 10242-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...