2-Hydroxyisonicotinic acid 98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
H64241.06
H64241.06EA
27.8
GBP
H64241.06
2-Hydroxyisonicotinic acid 98%
2-Hydroxyisonicotinic acid
Purity: 98%
CAS No.: 22282-72-0
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Physical Form: Solid
Beilstein Reference: 115859
MDL No.: MFCD01318566
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318566 CAS Number: 22282-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...