Bisoprolol hemifumarate 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H56014.MB H56014.MC
H56014.MBEA 180 GBP
H56014.MB H56014.MC
Bisoprolol hemifumarate 98%
Bisoprolol hemifumarate

Purity: 98% CAS No.: 104344-23-2 Molecular Formula: C18H31NO4.0.5C4H4O4 Formula Weight: 383.49 MDL No.: MFCD00792743

Formula: C₁₈H₃₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00792743
CAS Number: 104344-23-2
Merck Index: 14,1295

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR