5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(4-Chlorophenyl)-3-methyl-1H-pyrazol-5-amine
H51085.03EA
116
GBP
H51085.03
5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole 97%
5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole
Formula:
C₁₀H₁₀ClN₃ MW: 207.66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03478180 CAS Number: 40401-39-6 EINECS: 254-907-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...