Methyl-3-cyanocinnamate

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(3-Cyano-phenyl)-acrylic acid methyl ester

H50872.03EA 36.1 GBP
H50872.03
Methyl-3-cyanocinnamate
Methyl-3-cyanocinnamate
Formula: C₁₁H₉NO₂
MW: 187.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064268
CAS Number: 52116-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR