2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-6-sulphonyl chloride

H31670.03 H31670.06
H31670.03EA 56.4 GBP
H31670.03 H31670.06
2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride
2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride

CAS No.: 63758-12-3 Molecular Formula: C8H7ClO4S Formula Weight: 234.66 Melting Point: 65-67° UN No.: UN3261 MDL No.: MFCD02677681

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD02677681
CAS Number: 63758-12-3
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR