2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B20771.03 B20771.06
B20771.03EA 50.2 GBP
B20771.03 B20771.06
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 97%
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Purity: 97% CAS No.: 175201-51-1 Molecular Formula: C9H9N3O2 Formula Weight: 191.18 Melting Point: ca 250° dec. MDL No.: MFCD00067901

Formula: C₉H₉N₃O₂
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00067901
CAS Number: 175201-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR