Trifluoroacetaldehyde hydrate ≥75% in water, tech.

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Trifluoroacetaldehyde hydrate, 2,2,2-Trifluoro-1,1-ethanediol

A13234.18 A13234.09
A13234.18EA 272 GBP
A13234.18 A13234.09
Trifluoroacetaldehyde hydrate ≥75% in water, tech.
Trifluoroacetaldehyde hydrate

Purity: 75% CAS No.: 421-53-4 Molecular Formula: C2H3F3O2 Formula Weight: 116.04(anhy) Flash Point: None Boiling Point: 104-106° UN No.: UN1989 MDL No.: MFCD00167359

Formula: C₂H₃F₃O₂
MW: 116.039 g/mol
Boiling Pt: 104…106 °C
Density: 1.44
Storage Temperature: Ambient
MDL Number: MFCD00167359
CAS Number: 421-53-4
EINECS: 207-006-4
UN: 1989
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR