Benzyl-2,2,2-trichloroacetimidate 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Trichloroacetimidic Acid Benzyl Ester, Benzyl 2,2,2-trichloroacetimidate

A12387.06 A12387.14 A12387.18 A12387.30
A12387.06EA 24.5 GBP
A12387.06 A12387.14 A12387.18 A12387.30
Benzyl-2,2,2-trichloroacetimidate 98%
Benzyl-2,2,2-trichloroacetimidate

Purity: 98% CAS No.: 81927-55-1 Molecular Formula: C9H8Cl3NO Formula Weight: 252.53 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5450 Boiling Point: 110-114°/0.5mm Beilstein Reference: 2525375 MDL No.: MFCD00000805

Formula: C₉H₈Cl₃NO
MW: 252.52 g/mol
Boiling Pt: 110…114 °C (0.5 mmHg)
Density: 1.356
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00000805
CAS Number: 81927-55-1
EINECS: 617-271-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR