Mason's hygrometer
Supplier: Brannan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRNN13/222/3EA 88.1 GBP
BRNN13/222/3 BRNN13/222/2
Mason's hygrometer
Hygrometers Analogue Hygrometers
Mason's hygrometer are complete with wet and dry bulb and a large water reservoir. One thermometer bulb is in air and the other is wrapped in a wick kept moist from a water reservoir. The difference between the two readings is used to give an accurate calculation of humidity. These hygrometers are normally used in a stevenson screen.
  • Easy to read with red spirit scale
  • With polypropylene case