Power supplies, programmable, omniPAC MIDI
Supplier: Cleaver Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLEVCS-300VEA 0 GBP
CLEVCS-300V
Power supplies, programmable, omniPAC MIDI
Power Supplies
The omniPAC power supply can run as many as four units under a constant and programmable setting. A 3-digit LCD screen shows a particularly useful feature during extended runs when it is necessary to access the gel tank to monitor buffer levels and sample migration.
  • Stackable design
  • Automatic crossover
  • Dual voltage compatibility
Equipped with timer, alarm, pause/resume functionality, and enhanced in-built safety features.