Zinc iodide and starch solution analytical reagent, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
1.05445.0500EA
55.4
GBP
1.05445.0500
Zinc iodide and starch solution analytical reagent, Supelco®
Zinc iodide and starch solution
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.02 g/cm3 (20 °C) |
pH | 6.2 (H2O, 20 °C) |
Solubility | Soluble at 20 °C |
Storage temperature | +15...+25 °C |
Suitability as indicator | conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...