Dewar transportation flasks, in stainless steel
Supplier: Day-Impex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
479-0855EA 503 GBP
479-0855
Dewar transportation flasks, in stainless steel
Dewars and Cryogenic Vessels
These dewar transportation flasks are fitted into high quality stainless steel containers which results in them being rust free, anti-corrosive hygienic and strong. They have been designed with the focus on safety and practicality.
  • Rust-free, anti-corrosive, hygienic and strong
  • Stainless steel lid with self-contained insulation
  • Toggle clamps secure the lid to the body for safe transportation
Glass dewar fitted into high quality, stainless steel container.