Spherical ground joints, female
Supplier: Smith Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SMIT8SFS35EA 65.3 GBP
SMIT8SFS35
Spherical ground joints, female
Adapters Adapters, Spherical Ground Joints
Made of Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Through use of flexible spherical joint connections, apparatus can be assembled free from stresses.
Length: 100 mm