Anti-SRC Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP19615AEA 339 GBP
ABGEAP19615A
Anti-SRC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR