Anti-TRL4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK600-401-G52EA 690 GBP
ROCK600-401-G52
Anti-TRL4 Rabbit Polyclonal Antibody
Antibodies
Anti-TLR4 Antibody is suitable for use in WB, IHC, IF microscopy and Flow Cytometry. Expect a band approximately ~75-80kDa when tested against partial recombinant mouse TLR4 (extra-cellular portion plus His-tag).

Recommended Dilutions: Western Blot: 1-3µg/ml; Immunochemistry: 1:50

Type: Primary
Antigen: TRL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR