Anti-CD3e Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI138514EA 329 GBP
USBI138514
Anti-CD3e Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g

Type: Primary
Antigen: CD3e
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Chicken, Horse, Cat, Guinea pig, Hamster, Human, Macaque, Monkey, Mouse, Pig, Rabbit, Rat, Sheep,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR