Anti-Heat Shock Protein 90, alpha Mouse Monoclonal Antibody [clone: 10F737]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIH1834-55A3-200EA 564 GBP
USBIH1834-55A3-200
Anti-Heat Shock Protein 90, alpha Mouse Monoclonal Antibody [clone: 10F737]
Antibodies
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: Heat Shock Protein 90, alpha
Clonality: Monoclonal
Clone: 10F737
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR