Anti-Guanylin Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIG9805-03B-100EA 695 GBP
USBIG9805-03B-100 USBIG9805-03B-20
Anti-Guanylin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Guanylin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR