Anti-Insulin-Like Growth Factor Binding Protein 7 Rabbit Polyclonal Antibody (Biotin)

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBII7661-17A1-25EA 380 GBP
USBII7661-17A1-25 USBII7661-17A1-50
Anti-Insulin-Like Growth Factor Binding Protein 7 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: Insulin-Like Growth Factor Binding Protein 7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR