Anti-H-2K Mouse Monoclonal Antibody [clone: 5C10]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIH0007-20-1EA 804 GBP
USBIH0007-20-1
Anti-H-2K Mouse Monoclonal Antibody [clone: 5C10]
Antibodies
Type: Primary
Antigen: H-2K
Clonality: Monoclonal
Clone: 5C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR