Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 650)

Supplier: Agrisera
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AGRIAS122269EA 253 GBP
AGRIAS122269
Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR