Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Agrisera
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AGRIAS101177EA 240 GBP
AGRIAS101177
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Donkey
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR