Anti-IgM Antibody (HRP (Horseradish Peroxidase))

Supplier: Boster
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BSBTBA1077-0.25EA 0 GBP
BSBTBA1077-0.25
Anti-IgM Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR