Anti-Protein G Antibody (Biotin)

Supplier: Boster
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BSBTBA1026-0.5EA 0 GBP
BSBTBA1026-0.5
Anti-Protein G Antibody (Biotin)
Antibodies
Type: Tag
Antigen: Protein G
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR