Large capacity ovens
Supplier: Genlab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GENLLCO/30V/PDIGEA 0 GBP
GENLLCO/30V/PDIG
Large capacity ovens
Ovens Standard Ovens
These large capacity multi-purpose ovens offer a unique range of highly efficient,
reliable and accurate units suitable for much larger applications in drying, curing, general
laboratory and industrial uses.
  • Exterior is constructed from sheet steel finished in an easy clean powder coated paint
  • Interior chamber is made from mild steel coated with aluminium (CLAD) with a stainless steel chamber available as an option
  • Fitted with fixed shelf runners and removable chrome plated wire grid shelves
  • The vertical style units are fitted with single doors, the horizontal style units are fitted with twin doors
Heated by Incoloy sheathed elements positioned in the base fan duct for units up to 650 litres and in the side fan duct on units 650 litres and over.
All units have as standard a PID Microprocessor controller, with dual displays of set point and actual temperature that are auto-tuned to each individual unit to optimise the heat up, overshoot and control of temperature.
Delivery information: All units come with forced air fan circulation as standard.