Spill containment pallet
Supplier: Ecospill
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ESPAP3201420EA 1010 GBP
ESPAP3201420
Spill containment pallet
Spill Control Materials
Polyethylene single IBC with disposable tray.
  • Sturdy and corrosion resistant
  • Removable decking to assist cleaning
  • Forklift pockets for moving empty units