Viable microorganisms, Selectrol® discs
Supplier: TCS Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TCSBMM35EA 673 GBP
TCSBMM79 TCSBMM35 TCSBMD29 TCSBMM41T TCSBMD25 TCSBMD26 TCSBMD27 TCSBMD28 TCSBMD24 TCSBMD20 TCSBMM02 TCSBMM97 TCSBMM98 TCSBMM11 TCSBMM99 TCSBMD36 TCSBMM94 TCSBMD37 TCSBMM95 TCSBMD38 TCSBMD39 TCSBMM96 TCSBMD33 TCSBMD30 TCSBMD31 TCSBMD80 TCSBMD81 TCSBMD82 TCSBMM13 TCSBMD08 TCSBMD09 TCSBMD03 TCSBMD47 TCSBMD04 TCSBMD48 TCSBMD05 TCSBMD49 TCSBMM63 TCSBMD06 TCSBMD43 TCSBMD87 TCSBMD44 TCSBMD88 TCSBMD01 TCSBMD89 TCSBMD45 TCSBMD46 TCSBMD40 TCSBMD85 TCSBMD42 TCSBMD86 TCSBMD93 TCSBMM75 TCSBMD18 TCSBMD19 TCSBMM76 TCSBMM77 TCSBMD14 TCSBMD15 TCSBMD16 TCSBMM73 TCSBMD17 TCSBMD10 TCSBMD12
Viable microorganisms, Selectrol® discs
Microbiology Test Systems Microorganism Tests
Selectrol® discs are designed to be a convenient source of viable micro-organisms which can be used as controls for a variety of QC and other testing purposes. Selectrol® discs are convenient to store and versatile in their use. Selectrol® strains are presented as water-soluble freeze-dried discs with most strains being available in packs of 10 or 25 discs. Selectrol® discs are manufactured exclusively from NCTC® (National Collection of Type Cultures) and NCPF® (National Collection of Pathogenic Fungi) and are guaranteed first generation derivatives from the source strain. Selectrol® is versatile, and can be inoculated directly onto solid media or dissolved in an appropriate broth.
  • Fully traceable
  • Guaranteed first generation derivatives from original NCTC®/NCPF® cultures
  • Quality control micro-organisms of predictable biochemical reactions
  • Manufactured under licence from the Public Health England Culture Collections