Anti-IgY Goat Polyclonal Antibody (DyLight® 633)

Supplier: Immunoreagents
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IMMRIR1408EA 126 GBP
IMMRIR1408
Anti-IgY Goat Polyclonal Antibody (DyLight® 633)
Antibodies
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-Colour applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638nm/658nm Emission Colour = Red (Similar Dyes: Alexa Fluor 633).

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR