Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Immunoreagents
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IMMRIR1183EA 139 GBP
IMMRIR1183
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR