Anti-NCKX3 Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIN0559-06AEA 645 GBP
USBIN0559-06A
Anti-NCKX3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: NCKX3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR